The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_203507.3 | NP_987103.1 | 2 - 3 | 382 | 76 |
GenBank mRNA | AK004597.1 | - | 2 - 3 | 336 | 76 |
AK005797.1 | - | 2 - 3 | 367 | 76 | |
BB610203.1 | - | 2 - 3 | 383 | 76 | |
BC016198.1 | - | 2 - 3 | 318 | 76 |