Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Setd1b
Entrez Gene ID
208043
Gene Name
SET domain containing 1B
Gene Aliases
AA516740, BC035291, KMT2G, mKIAA1076
Chromosome Location
Chr.5: 123140787 - 123168634 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040398.2 NP_001035488.2 12 - 13 5508 73
GenBank mRNA AK122435.1 - 4 - 5 1843 73
BC035291.1 - 4 - 5 2010 73
BC038367.1 - 6 - 7 2586 73
BC040775.1 - 4 - 5 2010 73
BC041681.1 - 4 - 5 2029 73

View Full Product Details