The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013910.2 | NP_038938.1 | 3 - 4 | 569 | 70 |
GenBank mRNA | AK031626.1 | - | 3 - 4 | 534 | 70 |
AK129479.1 | - | 14 - 15 | 2006 | 70 | |
BC057622.1 | - | 3 - 4 | 564 | 70 | |
BC065090.1 | - | 14 - 15 | 2205 | 70 | |
BC068239.1 | - | 11 - 12 | 1759 | 70 | |
BU841175.1 | - | - | 601 | 70 |