The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027211.2 | NP_081487.1 | 2 - 3 | 142 | 90 |
GenBank mRNA | AJ306451.1 | - | 2 - 3 | 142 | 90 |
BB617542.1 | - | 2 - 3 | 139 | 90 | |
BC013521.1 | - | 2 - 3 | 112 | 90 |