The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008455.2 | NP_032481.2 | 12 - 13 | 1580 | 143 |
GenBank mRNA | BC026555.1 | - | 12 - 13 | 1506 | 143 |