The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145888.1 | NP_001139360.1 | 7 - 8 | 2631 | 137 |
NM_198644.2 | NP_941046.1 | 7 - 8 | 2610 | 137 | |
GenBank mRNA | BC053930.1 | - | 7 - 8 | 2601 | 137 |
EU221277.1 | - | 7 - 8 | 2461 | 137 |