Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ints1
Entrez Gene ID
68510
Gene Name
integrator complex subunit 1
Gene Aliases
1110015K06Rik
Chromosome Location
Chr.5: 139751282 - 139775701 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026748.2 NP_081024.3 19 - 20 2942 60
GenBank mRNA AB257854.1 - 19 - 20 2902 60
BC063266.1 - 19 - 20 2929 60
CB526682.1 - 6 - 7 661 60

View Full Product Details