Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kctd17
Entrez Gene ID
72844
Gene Name
potassium channel tetramerisation domain containing 17
Gene Aliases
2900008M13Rik, AA414907, AW742389, N28155
Chromosome Location
Chr.15: 78428559 - 78439303 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289671.1 NP_001276600.1 4 - 5 602 93
NM_001289672.1 NP_001276601.1 4 - 5 602 93
NM_001289673.1 NP_001276602.1 4 - 5 602 93
NR_110357.1 4 - 5 602 93
GenBank mRNA AK179755.1 - 3 - 4 290 93
BC144818.1 - 4 - 5 602 93
BC144820.1 - 4 - 5 602 93
BC145799.1 - 4 - 5 602 93

View Full Product Details