Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il2rb
Entrez Gene ID
16185
Gene Name
interleukin 2 receptor, beta chain
Gene Aliases
CD122, IL-15Rbeta, IL15Rbeta, Il-2/15Rbeta, Il-2Rbeta, p70
Chromosome Location
Chr.15: 78479256 - 78511627 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008368.4 NP_032394.1 6 - 7 692 68
GenBank mRNA AK017288.1 - 6 - 7 692 68
AK035289.1 - 6 - 7 645 68
AK157572.1 - 6 - 7 693 68
AK169481.1 - 6 - 7 673 68
BC026869.1 - 6 - 7 665 68
BC099616.1 - 6 - 7 666 68
M28052.1 - 6 - 7 649 68

View Full Product Details