Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rfx3
Entrez Gene ID
19726
Gene Name
regulatory factor X, 3 (influences HLA class II expression)
Gene Aliases
C230093O12Rik, MRFX3
Chromosome Location
Chr.19: 27761718 - 28011698 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166414.1 NP_001159886.1 4 - 5 380 91
NM_011265.3 NP_035395.2 3 - 4 421 91
GenBank mRNA AK030008.1 - 5 - 6 453 91
AK041120.1 - 4 - 5 481 91
AK140928.1 - 3 - 4 304 91
BC017598.1 - 3 - 4 395 91

View Full Product Details