Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Radil
Entrez Gene ID
231858
Gene Name
Ras association and DIL domains
Gene Aliases
AI536456, D930005D10Rik
Chromosome Location
Chr.5: 142484839 - 142551098 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289588.1 NP_001276517.1 9 - 10 2323 118
NM_178702.4 NP_848817.2 9 - 10 2323 118
GenBank mRNA AK052964.1 - 9 - 10 2234 118
AK079539.1 - 9 - 10 1758 118
AK083410.1 - 9 - 10 2341 118
AK154169.1 - 9 - 10 2268 118
AK162917.1 - 9 - 10 2304 118
AK173277.1 - 9 - 10 2287 118
BC056483.1 - 9 - 10 2275 118

View Full Product Details