The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081109.1 | NP_001074578.1 | 11 - 12 | 4256 | 86 |
GenBank mRNA | AB288872.1 | - | 11 - 12 | 4084 | 86 |
AK039738.1 | - | 1 - 2 | 232 | 86 | |
AK129279.2 | - | 1 - 2 | 2792 | 86 | |
BC058653.1 | - | 1 - 2 | 262 | 86 |