Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pvalb
Entrez Gene ID
19293
Gene Name
parvalbumin
Gene Aliases
PV, Parv, Pva
Chromosome Location
Chr.15: 78191117 - 78206351 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA S75909.1 - 1 - 2 70 75
S75910.1 - - 946 75
X59382.1 - 1 - 2 43 75

View Full Product Details