Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1qtnf6
Entrez Gene ID
72709
Gene Name
C1q and tumor necrosis factor related protein 6
Gene Aliases
2810036M19Rik, CTRP6
Chromosome Location
Chr.15: 78523346 - 78532191 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204152.1 NP_001191081.1 1 - 2 140 98
NM_001204153.1 NP_001191082.1 1 - 2 140 98
NM_028331.5 NP_082607.3 1 - 2 140 98
GenBank mRNA AK012868.1 - 1 - 2 83 98
AK051058.1 - 1 - 2 54 98
AK154407.1 - 1 - 2 117 98
BC025489.1 - 1 - 2 83 98
BC071187.1 - 1 - 2 107 98
DQ002399.1 - 1 - 2 106 98

View Full Product Details