Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ric1
Entrez Gene ID
226089
Gene Name
RAB6A GEF complex partner 1
Gene Aliases
C030046E11Rik, C130057E09Rik, Kiaa1432
Chromosome Location
Chr.19: 29522282 - 29605921 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081319.1 NP_001074788.1 18 - 19 2349 58
GenBank mRNA AK173171.1 - 17 - 18 1931 58
BC036137.1 - 2 - 3 145 58

View Full Product Details