The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172721.2 | NP_766309.2 | 10 - 11 | 1734 | 69 |
GenBank mRNA | AK031815.1 | - | 10 - 11 | 1728 | 69 |
AK034447.1 | - | 10 - 11 | 1662 | 69 | |
AK053794.1 | - | 10 - 11 | 1735 | 69 | |
AK134291.1 | - | 10 - 11 | 1742 | 69 | |
AK197164.1 | - | 1 - 2 | 66 | 69 | |
AK202489.1 | - | 1 - 2 | 65 | 69 | |
AK209645.1 | - | 1 - 2 | 65 | 69 | |
BC009095.1 | - | 10 - 11 | 1687 | 69 | |
BC024091.1 | - | 4 - 5 | 610 | 69 |