Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Taok3
Entrez Gene ID
330177
Gene Name
TAO kinase 3
Gene Aliases
2900006A08Rik, A130052D22, A430105I05Rik
Chromosome Location
Chr.5: 117120112 - 117275219 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081308.2 NP_001074777.1 14 - 15 1846 73
NM_001199685.1 NP_001186614.1 15 - 16 1954 73
NM_183306.2 NP_899129.2 15 - 16 1931 73
GenBank mRNA AK039665.1 - 2 - 3 504 73
AK042181.1 - 15 - 16 1854 73
AK155208.1 - 14 - 15 1842 73
BC158072.1 - 15 - 16 1896 73

View Full Product Details