Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Exoc6
Entrez Gene ID
107371
Gene Name
exocyst complex component 6
Gene Aliases
4833405E05Rik, AW413330, C430002C19, Sec15, Sec15l1, hbd, msec15
Chromosome Location
Chr.19: 37520874 - 37684052 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175353.2 NP_780562.2 17 - 18 1818 136
GenBank mRNA AK082881.1 - 17 - 18 1797 136
AK151524.1 - 17 - 18 1819 136
AK151838.1 - 17 - 18 1819 136
BC025616.1 - 4 - 5 392 136
BC026859.1 - 17 - 18 1771 136

View Full Product Details