Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Antxr2
Entrez Gene ID
71914
Gene Name
anthrax toxin receptor 2
Gene Aliases
2310046B19Rik, AW561899, CMG-2, CMG2, cI-35
Chromosome Location
Chr.5: 97884688 - 98030962 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133738.1 NP_598499.1 4 - 5 792 84
GenBank mRNA AK155460.1 - 4 - 5 875 84
AK166124.1 - - 564 84
AK170655.1 - 4 - 5 430 84
AK171292.1 - 4 - 5 467 84
AK190386.1 - 4 - 5 300 84
AK207219.1 - 4 - 5 300 84
BC076595.1 - 4 - 5 792 84

View Full Product Details