Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adrbk2
Entrez Gene ID
320129
Gene Name
adrenergic receptor kinase, beta 2
Gene Aliases
4833444A01Rik, AI851927, AW551196, Adrbk-2, Bark-2, GRK3
Chromosome Location
Chr.5: 112910478 - 113015951 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285806.1 NP_001272735.1 16 - 17 1492 80
NM_177078.4 NP_796052.2 16 - 17 1453 80
GenBank mRNA AK076533.1 - 16 - 17 1351 80
AK134672.1 - 16 - 17 1430 80
AK141186.1 - 16 - 17 1472 80

View Full Product Details