The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009745.2 | NP_033875.2 | 5 - 6 | 652 | 79 |
GenBank mRNA | AJ011145.1 | - | 5 - 6 | 532 | 79 |
AK131643.1 | - | 5 - 6 | 548 | 79 | |
AK155076.1 | - | 5 - 6 | 534 | 79 |