Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hps4
Entrez Gene ID
192232
Gene Name
Hermansky-Pudlak syndrome 4 homolog (human)
Gene Aliases
2010205O06Rik, AU040608, BLOC-3, C130020P05Rik, le, mKIAA1667
Chromosome Location
Chr.5: 112343056 - 112378424 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138646.3 NP_619587.3 9 - 10 1245 130
GenBank mRNA AK081489.1 - 9 - 10 1205 130
AK129418.1 - 8 - 9 1132 130
AK147954.1 - 8 - 9 1688 130
AK171871.1 - 9 - 10 1214 130
AY043415.1 - 8 - 9 765 130

View Full Product Details