Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Camkk2
Entrez Gene ID
207565
Gene Name
calcium/calmodulin-dependent protein kinase kinase 2, beta
Gene Aliases
6330570N16Rik, AW061083, mKIAA0787
Chromosome Location
Chr.5: 122718580 - 122779476 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199676.1 NP_001186605.1 16 - 17 1813 139
GenBank mRNA AF453383.1 - 15 - 16 1596 139
AK032070.1 - 16 - 17 1786 139
BC025895.1 - 4 - 5 267 139

View Full Product Details