Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc28
Entrez Gene ID
209683
Gene Name
tetratricopeptide repeat domain 28
Gene Aliases
2310015L07Rik, 6030435N04, AI428795, AI851761, BC002262, TPRBK
Chromosome Location
Chr.5: 110879784 - 111289780 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267622.1 NP_001254551.1 7 - 8 2867 104
GenBank mRNA AK122427.1 - 1 - 2 430 104
BC046779.1 - 1 - 2 531 104

View Full Product Details