Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnrc18
Entrez Gene ID
231861
Gene Name
trinucleotide repeat containing 18
Gene Aliases
BC025631, EG381742, Zfp469
Chromosome Location
Chr.5: 142724604 - 142822703 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122730.2 NP_001116202.2 20 - 21 6587 86
GenBank mRNA BC094365.1 - 1 - 2 116 86
CB244319.1 - 3 - 4 342 86

View Full Product Details