Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tfr2
Entrez Gene ID
50765
Gene Name
transferrin receptor 2
Gene Aliases
Trfr2
Chromosome Location
Chr.5: 137569822 - 137588081 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289507.1 NP_001276436.1 13 - 14 1800 120
NM_001289509.1 NP_001276438.1 15 - 16 1819 120
NM_001289511.1 NP_001276440.1 14 - 15 1677 120
NM_015799.4 NP_056614.3 15 - 16 1759 120
GenBank mRNA AF207741.1 - 13 - 14 1634 120
AF222895.1 - 14 - 15 1677 120
AK004848.2 - 15 - 16 1735 120
AK004965.1 - 15 - 16 1790 120
AK155526.1 - 14 - 15 1897 120
AK165320.1 - 13 - 14 1628 120
BC013654.1 - 13 - 14 1800 120

View Full Product Details