The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110828.2 | NP_001104298.1 | 1 - 2 | 284 | 97 |
NM_001110829.2 | NP_001104299.1 | 1 - 2 | 284 | 97 | |
NM_001110830.2 | NP_001104300.1 | 1 - 2 | 284 | 97 | |
NM_001286417.1 | NP_001273346.1 | 1 - 2 | 284 | 97 | |
NM_001286418.1 | NP_001273347.1 | 1 - 2 | 284 | 97 | |
NM_001286419.1 | NP_001273348.1 | 1 - 2 | 284 | 97 | |
GenBank mRNA | AF229055.1 | - | 1 - 2 | 227 | 97 |
BC002124.1 | - | 1 - 2 | 122 | 97 | |
BY247199.1 | - | 1 - 2 | 286 | 97 | |
BY254584.1 | - | 1 - 2 | 285 | 97 |