Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rph3a
Entrez Gene ID
19894
Gene Name
rabphilin 3A
Gene Aliases
2900002P20Rik, AU022689, AW108370
Chromosome Location
Chr.5: 120940499 - 121010102 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB112924.1 - 1 - 2 110 74
AU067470.1 - 1 - 2 298 74
BC042585.1 - 1 - 2 278 74
CB526789.1 - 2 - 3 465 74
D29965.1 - 1 - 2 89 74

View Full Product Details