The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010596.2 | NP_034726.2 | 1 - 2 | 1054 | 134 |
GenBank mRNA | AY779767.1 | - | 1 - 2 | 788 | 134 |