Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ms4a18
Entrez Gene ID
76002
Gene Name
membrane-spanning 4-domains, subfamily A, member 18
Gene Aliases
5033428B15Rik
Chromosome Location
Chr.19: 10997024 - 11018026 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251849.1 NP_001238778.1 2 - 3 544 129

View Full Product Details