Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ms4a10
Entrez Gene ID
69826
Gene Name
membrane-spanning 4-domains, subfamily A, member 10
Gene Aliases
2010001N17Rik
Chromosome Location
Chr.19: 10962289 - 10974725 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023529.2 NP_076018.2 7 - 8 760 67
GenBank mRNA AF237918.1 - 7 - 8 760 67
AK008019.1 - 5 - 6 619 67
BC144935.1 - 7 - 8 757 67
BC150675.1 - 7 - 8 757 67

View Full Product Details