Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccnj
Entrez Gene ID
240665
Gene Name
cyclin J
Gene Aliases
D430039C20Rik
Chromosome Location
Chr.19: 40831279 - 40848572 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172839.4 NP_766427.1 4 - 5 790 119
GenBank mRNA AK052506.1 - 4 - 5 791 119
AK157841.1 - 4 - 5 784 119
BC050857.1 - 1 - 2 241 119
BC120921.2 - 4 - 5 772 119
BC120922.1 - 4 - 5 772 119

View Full Product Details