The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133352.2 | NP_579930.1 | 2 - 3 | 449 | 125 |
GenBank mRNA | AF269151.1 | - | 2 - 3 | 381 | 125 |
AK075807.1 | - | 2 - 3 | 417 | 125 | |
AK147380.1 | - | 2 - 3 | 444 | 125 | |
AK147629.1 | - | 2 - 3 | 443 | 125 | |
AK220279.1 | - | 2 - 3 | 425 | 125 | |
BC004799.1 | - | 1 - 2 | 78 | 125 | |
CA529050.1 | - | 2 - 3 | 451 | 125 |