Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ubxn8
Entrez Gene ID
108159
Gene Name
UBX domain protein 8
Gene Aliases
D0H8S2298E, Rep-8, Rep8h, Ubxd6
Chromosome Location
Chr.8: 33619586 - 33643762 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178648.2 NP_848763.2 6 - 7 624 88
GenBank mRNA AK011496.1 - 6 - 7 612 88
AK016964.1 - 5 - 6 496 88
AK132565.1 - 6 - 7 617 88
AK161397.1 - 6 - 7 1019 88
BC024492.1 - 6 - 7 618 88
D88447.1 - 6 - 7 607 88

View Full Product Details