The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144549.4 | NP_653132.1 | 1 - 2 | 873 | 79 |
GenBank mRNA | AF358866.1 | - | 1 - 2 | 456 | 79 |
AK028626.1 | - | 1 - 2 | 874 | 79 | |
AK033358.1 | - | 1 - 2 | 875 | 79 | |
AK040738.1 | - | 1 - 2 | 874 | 79 | |
AK041212.1 | - | 1 - 2 | 874 | 79 | |
AK044537.1 | - | 1 - 2 | 1182 | 79 | |
AK080228.1 | - | 1 - 2 | 847 | 79 | |
AK162876.1 | - | 1 - 2 | 874 | 79 | |
BC006800.1 | - | 1 - 2 | 457 | 79 |