The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177464.4 | NP_803415.1 | 3 - 4 | 2015 | 87 |
GenBank mRNA | AK082926.1 | - | 3 - 4 | 2016 | 87 |
AK147893.1 | - | 3 - 4 | 1999 | 87 | |
BC038935.1 | - | 3 - 4 | 1991 | 87 |