The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023740.2 | NP_076229.2 | 1 - 2 | 193 | 97 |
GenBank mRNA | AF176814.1 | - | 1 - 2 | 166 | 97 |
AK151505.1 | - | 1 - 2 | 175 | 97 | |
AK168042.1 | - | 1 - 2 | 168 | 97 | |
BB852670.1 | - | 1 - 2 | 196 | 97 | |
BC010835.1 | - | 1 - 2 | 157 | 97 | |
CK619221.1 | - | 1 - 2 | 180 | 97 |