Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Arhgap19
- Entrez Gene ID
-
71085
- Gene Name
- Rho GTPase activating protein 19
- Gene Aliases
-
4933411B03Rik, AI120128, AW546831, mFLJ00368
- Chromosome Location
-
Chr.19: 41766588 - 41802084 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK016763.1 |
- |
6 - 7 |
955 |
157 |
View Full Product Details