Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fgl1
Entrez Gene ID
234199
Gene Name
fibrinogen-like protein 1
Gene Aliases
Mfire1
Chromosome Location
Chr.8: 41191434 - 41217978 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145594.2 NP_663569.2 4 - 5 496 95
GenBank mRNA AF478470.1 - 4 - 5 506 95
BC021946.1 - 4 - 5 464 95
BC029734.1 - 4 - 5 473 95

View Full Product Details