Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dgkq
Entrez Gene ID
110524
Gene Name
diacylglycerol kinase, theta
Gene Aliases
110kDa, DAGK, DAGK7, Dagk4, Dgkd
Chromosome Location
Chr.5: 108647044 - 108660828 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199011.1 NP_950176.1 10 - 11 1356 74
GenBank mRNA AK043933.1 - - 1206 74
AK048764.1 - 10 - 11 1398 74
AK170840.1 - 9 - 10 1452 74
BC062929.1 - 10 - 11 1356 74
BC066012.1 - 10 - 11 1333 74

View Full Product Details