Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dgkq
Entrez Gene ID
110524
Gene Name
diacylglycerol kinase, theta
Gene Aliases
110kDa, DAGK, DAGK7, Dagk4, Dgkd
Chromosome Location
Chr.5: 108647044 - 108660828 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199011.1 NP_950176.1 6 - 7 872 64
GenBank mRNA AK043933.1 - - 722 64
AK048764.1 - 6 - 7 914 64
AK132151.1 - 6 - 7 869 64
BC062929.1 - 6 - 7 872 64
BC066012.1 - 6 - 7 849 64

View Full Product Details