Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc18
Entrez Gene ID
73254
Gene Name
coiled-coil domain containing 18
Gene Aliases
1700021E15Rik, 4932411G06Rik, NY-SAR-41
Chromosome Location
Chr.5: 108132619 - 108232951 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028481.1 NP_082757.1 27 - 28 3959 101
GenBank mRNA AK016519.1 - 27 - 28 3954 101
AK045927.1 - - 100
BC082598.1 - 27 - 28 3959 101

View Full Product Details