The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028481.1 | NP_082757.1 | 28 - 29 | 4439 | 131 |
GenBank mRNA | AK016519.1 | - | 28 - 29 | 4434 | 131 |
BC082598.1 | - | 28 - 29 | 4439 | 131 |