The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_054071.2 | NP_473412.1 | 3 - 4 | 802 | 141 |
GenBank mRNA | AF321300.1 | - | 3 - 4 | 529 | 141 |
AJ293947.1 | - | 2 - 3 | 435 | 141 | |
BC058745.1 | - | 2 - 3 | 404 | 141 |