Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pigg
Entrez Gene ID
433931
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class G
Gene Aliases
Gpi7
Chromosome Location
Chr.5: 108312783 - 108349359 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081234.1 NP_001074703.1 10 - 11 2241 89
GenBank mRNA BC051059.1 - 4 - 5 1122 89

View Full Product Details