The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018801.3 | NP_061271.1 | 9 - 9 | 6271 | 158 |
NM_173067.3 | NP_775090.1 | 10 - 10 | 6403 | 158 | |
NM_173068.2 | NP_775091.2 | 11 - 11 | 6763 | 158 | |
GenBank mRNA | AK080792.1 | - | 1 - 1 | 1113 | 158 |