Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fndc7
Entrez Gene ID
320181
Gene Name
fibronectin type III domain containing 7
Gene Aliases
E230011A21Rik
Chromosome Location
Chr.3: 108853678 - 108890008 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165965.1 NP_001159437.1 2 - 3 387 119
NM_177091.5 NP_796065.2 2 - 3 387 119
GenBank mRNA AK054241.1 - 2 - 3 387 119
BC111898.1 - 2 - 3 194 119

View Full Product Details