Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tctn2
Entrez Gene ID
67978
Gene Name
tectonic family member 2
Gene Aliases
4432405B04Rik, Tect2
Chromosome Location
Chr.5: 124598749 - 124627738 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026486.3 NP_080762.1 1 - 2 207 120
GenBank mRNA AK012720.1 - 1 - 2 180 120
AK014476.1 - 1 - 2 173 120
AK031829.1 - 1 - 2 208 120
AK087856.1 - 1 - 2 165 120
BC058375.1 - 1 - 2 159 120
DQ278869.1 - 1 - 2 205 120

View Full Product Details