The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019875.2 | NP_063928.2 | 11 - 12 | 2274 | 64 |
GenBank mRNA | AB093298.1 | - | 11 - 12 | 2256 | 64 |
AF216495.1 | - | 11 - 12 | 2267 | 64 | |
AK044140.1 | - | 11 - 12 | 2252 | 64 | |
BC053014.1 | - | 11 - 12 | 2227 | 64 |