Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abcb9
Entrez Gene ID
56325
Gene Name
ATP-binding cassette, sub-family B (MDR/TAP), member 9
Gene Aliases
TAPL, mKIAA1520
Chromosome Location
Chr.5: 124061857 - 124095966 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019875.2 NP_063928.2 1 - 2 158 72
GenBank mRNA AB093298.1 - 1 - 2 140 72
AF216495.1 - 1 - 2 151 72
AK044140.1 - 1 - 2 136 72
BC026455.1 - 1 - 2 146 72
BC053014.1 - 1 - 2 111 72
BY215875.1 - 1 - 2 159 72

View Full Product Details